Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200014844.mm.1 |
NM_175550.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014844.mm.1 |
NM_175550.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014844.mm.1 |
TC0200001904.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014842.mm.1 |
NM_175550.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014842.mm.1 |
NM_175550.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014842.mm.1 |
TC0200001904.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014845.mm.1 |
NM_175550.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014845.mm.1 |
NM_175550.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014845.mm.1 |
TC0200001904.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014840.mm.1 |
NM_175550.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014840.mm.1 |
NM_175550.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014840.mm.1 |
TC0200001904.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014843.mm.1 |
NM_175550.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014843.mm.1 |
NM_175550.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014843.mm.1 |
TC0200001904.mm.1 |
MTA TC |