Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phospho2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028521.2 | NP_082797.1 | 1 - 2 | 213 | 107 | |
GenBank mRNA | AK006724.1 | - | 1 - 2 | 214 | 107 | |
AK010147.1 | - | 239 | 107 | |||
AK159551.1 | - | 1 - 2 | 261 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |