Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK004314.1 | - | 3 - 4 | 307 | 74 | |
AK050359.1 | - | 7 - 8 | 772 | 74 | ||
AK173334.1 | - | 5 - 6 | 513 | 74 | ||
BC085286.1 | - | 8 - 9 | 898 | 74 | ||
BC157930.1 | - | 15 - 16 | 2203 | 74 | ||
BC172022.1 | - | 15 - 16 | 2204 | 74 | ||
BC172146.1 | - | 35 - 36 | 4770 | 74 | ||
DQ924536.1 | - | 35 - 36 | 5067 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |