Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200031791.mm.1 |
NM_172664.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031791.mm.1 |
NM_172664.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031791.mm.1 |
TC0200003712.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031789.mm.1 |
NM_172664.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031789.mm.1 |
NM_172664.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031789.mm.1 |
TC0200003712.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031793.mm.1 |
NM_172664.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031793.mm.1 |
NM_172664.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031793.mm.1 |
TC0200003712.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031787.mm.1 |
NM_172664.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031787.mm.1 |
NM_172664.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031787.mm.1 |
TC0200003712.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031792.mm.1 |
NM_172664.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031792.mm.1 |
NM_172664.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031792.mm.1 |
TC0200003712.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031794.mm.1 |
NM_172664.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031794.mm.1 |
NM_172664.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031794.mm.1 |
TC0200003712.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031796.mm.1 |
NM_172664.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031796.mm.1 |
NM_172664.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031796.mm.1 |
TC0200003712.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031797.mm.1 |
NM_172664.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031797.mm.1 |
NM_172664.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031797.mm.1 |
TC0200003712.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031788.mm.1 |
NM_172664.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031788.mm.1 |
NM_172664.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031788.mm.1 |
TC0200003712.mm.1 |
MTA TC |