Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hhipl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044380.1 | NP_001037845.1 | 3 - 4 | 1210 | 64 | |
GenBank mRNA | AK129450.1 | - | 605 | 64 | ||
AK138646.1 | - | 3 - 4 | 934 | 64 | ||
AK147996.1 | - | 877 | 64 | |||
AK166269.1 | - | 1106 | 64 | |||
BC057108.1 | - | 203 | 64 | |||
BC113163.1 | - | 3 - 4 | 1210 | 40060054 | 64 | |
BC113773.1 | - | 3 - 4 | 1206 | 40060051 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |