Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glp2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175681.3 | NP_783612.2 | 10 - 11 | 1317 | 102 | |
GenBank mRNA | AK035702.1 | - | 712 | 102 | ||
AY605231.1 | - | 10 - 11 | 1036 | 102 | ||
BC044746.2 | - | 10 - 11 | 1317 | 5363416 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |