Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013013.2 | NP_001013031.2 | 4 - 5 | 618 | 100 | |
GenBank mRNA | AI391290.1 | - | 1 - 2 | 107 | 100 | |
BC039565.1 | - | 4 - 5 | 509 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |