Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153501.2 | NP_705721.2 | 2 - 3 | 667 | 159 | |
GenBank mRNA | AK016755.1 | - | 4 - 5 | 690 | 159 | |
AK082821.1 | - | 2 - 3 | 673 | 159 | ||
AK154080.1 | - | 2 - 3 | 631 | 159 | ||
AK159175.1 | - | 2 - 3 | 658 | 160 | ||
AK167861.1 | - | 2 - 3 | 375 | 159 | ||
BC106184.1 | - | 2 - 3 | 621 | 30241166 | 159 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |