Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200039607.mm.1 |
NM_001255993.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039607.mm.1 |
NM_001255993.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039607.mm.1 |
NM_001255994.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039607.mm.1 |
NM_001255994.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039607.mm.1 |
TC0200004631.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200039618.mm.1 |
NM_001146084.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039618.mm.1 |
NM_001146084.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039618.mm.1 |
NM_001255993.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039618.mm.1 |
NM_001255993.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039618.mm.1 |
NM_001255994.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039618.mm.1 |
NM_001255994.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039618.mm.1 |
NM_080562.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039618.mm.1 |
NM_080562.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039618.mm.1 |
NM_198176.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039618.mm.1 |
NM_198176.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039618.mm.1 |
TC0200004631.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200039619.mm.1 |
NM_001146084.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039619.mm.1 |
NM_001146084.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039619.mm.1 |
NM_001255993.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039619.mm.1 |
NM_001255993.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039619.mm.1 |
NM_001255994.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039619.mm.1 |
NM_001255994.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039619.mm.1 |
NM_080562.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039619.mm.1 |
NM_080562.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039619.mm.1 |
NM_198176.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039619.mm.1 |
NM_198176.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039619.mm.1 |
TC0200004631.mm.1 |
MTA TC |