Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025922.2 | NP_080198.2 | 2 - 3 | 156 | 58 | |
GenBank mRNA | AB100501.1 | - | 2 - 3 | 168 | 58 | |
AK009248.1 | - | 2 - 3 | 169 | 58 | ||
AK138101.1 | - | 2 - 3 | 196 | 58 | ||
AK139770.1 | - | 2 - 3 | 154 | 58 | ||
AK153806.1 | - | 2 - 3 | 196 | 58 | ||
AK153904.1 | - | 2 - 3 | 175 | 58 | ||
BC026508.1 | - | 2 - 3 | 156 | 4512398 | 58 | |
BC094466.1 | - | 2 - 3 | 144 | 4936811 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |