Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21633476 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21633478 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P130332 |
BC011103.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P130332 |
BC011103.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
491663 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
311172 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572495 |
AK080223.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572495 |
XM_001472981.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572495 |
XM_001473006.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572495 |
XM_896373.2 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
478750 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
943167 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
379619 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
182478 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
368786 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
433575 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
301697 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
742121 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
57998 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
2365 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P122562 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P104858 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P130332 |
BC011103.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P130332 |
BC011103.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633376 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633339 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633362 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633324 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633351 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633361 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633363 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633370 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633352 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633377 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633366 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633334 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633343 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633355 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633321 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633347 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633333 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633346 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633348 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633337 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633359 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633378 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633375 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633373 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633367 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633374 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633372 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633358 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633368 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633344 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633365 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633328 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633357 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633369 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633326 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633335 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633331 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21633329 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04693977 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P122562 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P104858 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P130332 |
BC011103.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P130332 |
BC011103.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P122928 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P122562 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P104858 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01524579 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01524557 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01524575 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01524597 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01524574 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01524559 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21633478 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21633476 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633510 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633472 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633368 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633394 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633400 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633532 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633439 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633504 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633546 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633355 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633318 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633481 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633452 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633421 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633373 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633553 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633359 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633390 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633502 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633378 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633485 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633458 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633433 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633410 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633347 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633531 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633326 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633526 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633376 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633414 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633438 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633406 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633488 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633428 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633370 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633492 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633436 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633528 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633395 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633536 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633392 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633507 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633520 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633349 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633346 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633331 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633442 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633351 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633443 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633367 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633397 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633496 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633501 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633429 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633328 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633432 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633498 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633365 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633377 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633476 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633417 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633338 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633375 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633450 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633382 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633348 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633343 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633462 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633480 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633508 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633426 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633387 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633404 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633379 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633333 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633386 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633497 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633547 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633358 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633463 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633366 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633415 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633408 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633541 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633535 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633516 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633361 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633324 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633457 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633495 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633468 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633445 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633505 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633479 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633393 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633506 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633363 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633469 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633490 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633402 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633335 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633329 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633551 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633539 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633412 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633522 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633544 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633357 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633512 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633530 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633447 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633550 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633454 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21633403 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633368 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633344 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633319 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633402 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01294303 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01524590 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01524599 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01602943 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633450 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01602951 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633362 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633382 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633438 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633443 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01602953 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01602938 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633387 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633442 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01236109 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633334 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633448 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633453 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633367 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02238328 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01971364 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633452 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02238332 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633407 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01294298 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01524587 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633351 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01602956 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633343 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633447 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633328 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01221768 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633451 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01524565 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633361 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02144728 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633324 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01236110 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633359 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02146452 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633393 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02238329 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01524569 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633375 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01602946 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633330 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633357 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01524592 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633355 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633390 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01524598 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633318 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01602949 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633392 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01602957 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633331 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633439 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01524586 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633317 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633445 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01316389 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633365 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633338 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633322 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633346 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01602937 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21633321 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01524570 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01602947 |
- |
- |