Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc25b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111075.4 | NP_001104545.1 | 11 - 12 | 1405 | 75 | |
NM_023117.4 | NP_075606.1 | 12 - 13 | 1483 | 75 | ||
GenBank mRNA | AK004836.1 | - | 11 - 12 | 1362 | 75 | |
AK043730.1 | - | 11 - 12 | 1396 | 75 | ||
AK153903.1 | - | 12 - 13 | 1484 | 75 | ||
BC002287.1 | - | 3 - 4 | 270 | 75 | ||
BC057568.1 | - | 11 - 12 | 1276 | 6401489 | 75 | |
S93521.1 | - | 12 - 13 | 1352 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |