Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291434.1 | NP_001278363.1 | 14 - 15 | 1760 | 74 | |
NM_011102.4 | NP_035232.1 | 15 - 16 | 1913 | 74 | ||
GenBank mRNA | AK144419.1 | - | 15 - 16 | 1915 | 74 | |
BC111807.1 | - | 14 - 15 | 1647 | 40039740 | 74 | |
L28035.1 | - | 15 - 16 | 1991 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |