Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007387.2 | NP_031413.1 | 5 - 6 | 574 | 65 | |
GenBank mRNA | AK089401.1 | - | 5 - 6 | 565 | 65 | |
AK128963.1 | - | 5 - 6 | 567 | 65 | ||
AK133756.1 | - | 5 - 6 | 570 | 65 | ||
AK154269.1 | - | 5 - 6 | 570 | 65 | ||
AK170658.1 | - | 5 - 6 | 574 | 65 | ||
BC023343.1 | - | 5 - 6 | 556 | 5136781 | 65 | |
X57199.1 | - | 5 - 6 | 545 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |