Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290807.1 | NP_001277736.1 | 4 - 5 | 358 | 69 | |
NM_001290808.1 | NP_001277737.1 | 3 - 4 | 388 | 69 | ||
NM_025475.3 | NP_079751.1 | 4 - 5 | 458 | 69 | ||
GenBank mRNA | AK010445.1 | - | 4 - 5 | 405 | 69 | |
AK012086.1 | - | 4 - 5 | 457 | 69 | ||
AK012779.1 | - | 3 - 4 | 388 | 69 | ||
AK012960.1 | - | 4 - 5 | 405 | 69 | ||
AK132300.1 | - | 4 - 5 | 414 | 69 | ||
BC086677.1 | - | 4 - 5 | 396 | 30660268 | 69 | |
BY711279.1 | - | 4 - 5 | 460 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |