Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199004.1 | NP_001185933.1 | 11 - 12 | 2307 | 100 | |
NM_013747.4 | NP_038775.1 | 11 - 12 | 2149 | 100 | ||
GenBank mRNA | AB016784.1 | - | 11 - 12 | 2308 | 100 | |
AF026274.1 | - | 11 - 12 | 2139 | 100 | ||
AK138455.1 | - | 11 - 12 | 2308 | 100 | ||
AK152533.1 | - | 11 - 12 | 2157 | 100 | ||
AK153010.1 | - | 11 - 12 | 2157 | 100 | ||
AK168765.1 | - | 11 - 12 | 2153 | 100 | ||
BC016883.1 | - | 11 - 12 | 2877 | 4235952 | 100 | |
BC086782.1 | - | 11 - 12 | 2149 | 6813878 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |