Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019488.4 | NP_062361.1 | 3 - 4 | 468 | 102 | |
GenBank mRNA | AF232061.2 | - | 3 - 4 | 425 | 102 | |
AJ245936.1 | - | 3 - 4 | 445 | 102 | ||
AK081806.1 | - | 3 - 4 | 485 | 102 | ||
AK141193.1 | - | 2 - 3 | 634 | 102 | ||
AK155792.1 | - | 3 - 4 | 497 | 103 | ||
AK170319.1 | - | 3 - 4 | 481 | 102 | ||
AY856043.1 | - | 3 - 4 | 429 | 102 | ||
AY856044.1 | - | 3 - 4 | 429 | 102 | ||
AY856045.1 | - | 3 - 4 | 429 | 102 | ||
BC090993.1 | - | 3 - 4 | 468 | 6751763 | 102 | |
Y17802.1 | - | 3 - 4 | 450 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |