Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026790.2 | NP_081066.2 | 4 - 5 | 571 | 112 | |
GenBank mRNA | AK075797.2 | - | 4 - 5 | 510 | 112 | |
AK169926.1 | - | 4 - 5 | 501 | 112 | ||
BC128276.1 | - | 4 - 5 | 470 | 40091489 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |