Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026790.2 | NP_081066.2 | 5 - 6 | 600 | 58 | |
NM_194067.2 | NP_919043.1 | 4 - 5 | 465 | 58 | ||
GenBank mRNA | AJ566111.1 | - | 5 - 6 | 578 | 58 | |
AJ566113.1 | - | 4 - 5 | 443 | 58 | ||
AK075797.2 | - | 5 - 6 | 539 | 58 | ||
AK146231.1 | - | 4 - 5 | 442 | 58 | ||
AK169926.1 | - | 5 - 6 | 530 | 58 | ||
BC128276.1 | - | 5 - 6 | 499 | 40091489 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |