Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ell3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145973.2 | NP_666085.2 | 2 - 3 | 241 | 55 | |
GenBank mRNA | AK139676.1 | - | 3 - 4 | 667 | 55 | |
BC045151.1 | - | 2 - 3 | 241 | 6313358 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |