Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029959.2 | NP_084235.1 | 5 - 6 | 522 | 77 | |
GenBank mRNA | AF435738.1 | - | 5 - 6 | 522 | 77 | |
AK020305.1 | - | 5 - 6 | 524 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |