Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133835.2 | NP_598596.2 | 6 - 7 | 873 | 114 | |
GenBank mRNA | AK157857.1 | - | 6 - 7 | 707 | 114 | |
AK161969.1 | - | 1 - 2 | 76 | 114 | ||
AK177482.1 | - | 1 - 2 | 56 | 114 | ||
AK186117.1 | - | 1 - 2 | 56 | 114 | ||
AK187016.1 | - | 1 - 2 | 56 | 114 | ||
AK209158.1 | - | 1 - 2 | 49 | 114 | ||
BC021811.1 | - | 6 - 7 | 824 | 3662474 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |