Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290437.1 | NP_001277366.1 | 3 - 4 | 1293 | 105 | |
NM_033134.3 | NP_149125.1 | 2 - 3 | 1577 | 105 | ||
GenBank mRNA | AF226683.1 | - | 2 - 3 | 1539 | 105 | |
AK047245.1 | - | 2 - 3 | 1562 | 105 | ||
AK080075.1 | - | 2 - 3 | 1559 | 105 | ||
AK085439.1 | - | 3 - 4 | 1297 | 105 | ||
AK154097.1 | - | 2 - 3 | 1578 | 105 | ||
AK158135.1 | - | 2 - 3 | 1562 | 105 | ||
AK170786.1 | - | 2 - 3 | 1534 | 105 | ||
AK170812.1 | - | 2 - 3 | 1548 | 105 | ||
BC052717.1 | - | 2 - 3 | 1546 | 6837339 | 105 | |
BC080295.1 | - | 2 - 3 | 1542 | 30354397 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |