Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290437.1 | NP_001277366.1 | 7 - 8 | 1739 | 71 | |
NM_033134.3 | NP_149125.1 | 6 - 7 | 2023 | 71 | ||
NR_110957.1 | 6 - 7 | 2110 | 71 | |||
GenBank mRNA | AF226683.1 | - | 6 - 7 | 1985 | 71 | |
AK047245.1 | - | 5 - 6 | 1888 | 71 | ||
AK080075.1 | - | 6 - 7 | 2005 | 71 | ||
AK085439.1 | - | 7 - 8 | 1744 | 71 | ||
AK154097.1 | - | 6 - 7 | 2024 | 71 | ||
AK158135.1 | - | 6 - 7 | 2008 | 71 | ||
AK170174.1 | - | 7 - 8 | 2029 | 71 | ||
AK170786.1 | - | 6 - 7 | 1980 | 71 | ||
AK170812.1 | - | 6 - 7 | 1994 | 71 | ||
BC052717.1 | - | 6 - 7 | 1992 | 6837339 | 71 | |
BC057566.1 | - | 6 - 7 | 2075 | 71 | ||
BC080295.1 | - | 6 - 7 | 1988 | 30354397 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |