Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276448.1 | NP_001263377.1 | 6 - 7 | 658 | 73 | |
GenBank mRNA | AK033743.1 | - | 4 - 5 | 1185 | 73 | |
FJ465144.1 | - | 6 - 7 | 635 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons |