Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009728.2 | NP_033858.2 | 6 - 7 | 1370 | 160 | |
GenBank mRNA | AF011337.1 | - | 6 - 7 | 1073 | 160 | |
AF156549.1 | - | 6 - 7 | 1240 | 160 | ||
AK132174.1 | - | 6 - 7 | 1327 | 160 | ||
AK172149.1 | - | 6 - 7 | 1291 | 160 | ||
AK172972.1 | - | 6 - 7 | 1156 | 160 | ||
BC138356.1 | - | 6 - 7 | 1174 | 8861376 | 160 | |
BC138357.1 | - | 6 - 7 | 1174 | 8861377 | 160 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |