Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009728.2 | NP_033858.2 | 7 - 8 | 1631 | 70 | |
GenBank mRNA | AF156549.1 | - | 7 - 8 | 1501 | 70 | |
AK132174.1 | - | 7 - 8 | 1588 | 70 | ||
AK172149.1 | - | 7 - 8 | 1552 | 70 | ||
AK172972.1 | - | 7 - 8 | 1417 | 70 | ||
BC039884.1 | - | 1 - 2 | 218 | 70 | ||
BC138356.1 | - | 7 - 8 | 1435 | 8861376 | 70 | |
BC138357.1 | - | 7 - 8 | 1435 | 8861377 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |