Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009728.2 | NP_033858.2 | 8 - 9 | 1847 | 78 | |
GenBank mRNA | AF156549.1 | - | 8 - 9 | 1717 | 78 | |
AK132174.1 | - | 8 - 9 | 1804 | 78 | ||
AK172149.1 | - | 8 - 9 | 1768 | 78 | ||
AK172972.1 | - | 8 - 9 | 1633 | 78 | ||
BC039884.1 | - | 2 - 3 | 434 | 78 | ||
BC138356.1 | - | 8 - 9 | 1651 | 8861376 | 78 | |
BC138357.1 | - | 8 - 9 | 1651 | 8861377 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |