Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp6r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172894.2 | NP_766482.2 | 18 - 19 | 2591 | 58 | |
GenBank mRNA | AK075825.1 | - | 2 - 3 | 303 | 58 | |
AK122450.1 | - | 14 - 15 | 3695 | 58 | ||
BC053076.1 | - | 18 - 19 | 2591 | 5698212 | 58 | |
BC141039.1 | - | 17 - 18 | 2138 | 9055651 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |