Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011618.2 | NP_035748.1 | 6 - 7 | 181 | 84 | |
GenBank mRNA | AK160226.1 | - | 6 - 7 | 182 | 84 | |
BC138871.1 | - | 6 - 7 | 161 | 8861893 | 84 | |
U92883.1 | - | 6 - 7 | 165 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |