Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200030455.mm.1 |
NM_001033369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030455.mm.1 |
NM_001033369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030455.mm.1 |
NM_001111030.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030455.mm.1 |
NM_001111030.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030455.mm.1 |
TC0200003576.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200030456.mm.1 |
NM_001033369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030456.mm.1 |
NM_001033369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030456.mm.1 |
NM_001111030.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030456.mm.1 |
NM_001111030.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030456.mm.1 |
TC0200003576.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200030459.mm.1 |
NM_001111030.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030459.mm.1 |
NM_001111030.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030459.mm.1 |
TC0200003576.mm.1 |
MTA TC |