Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acvr1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033369.3 | NP_001028541.1 | 7 - 8 | 1071 | 59 | |
NM_001111030.1 | NP_001104500.1 | 8 - 9 | 1438 | 59 | ||
GenBank mRNA | AK049089.1 | - | 7 - 8 | 1072 | 59 | |
AK142396.1 | - | 8 - 9 | 1442 | 59 | ||
BC028780.1 | - | 4 - 5 | 456 | 59 | ||
BC145224.1 | - | 7 - 8 | 1234 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |