Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289660.1 | NP_001276589.1 | 2 - 3 | 267 | 106 | |
NM_029692.3 | NP_083968.1 | 1 - 2 | 246 | 106 | ||
GenBank mRNA | AK044005.1 | - | 2 - 3 | 215 | 106 | |
AK149444.1 | - | 1 - 2 | 244 | 106 | ||
AY152393.1 | - | 1 - 2 | 245 | 106 | ||
BY133124.1 | - | 1 - 2 | 247 | 106 | ||
CJ130127.1 | - | 2 - 3 | 268 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus 1700 Probe spans exons |