Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acvr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007396.4 | NP_031422.3 | 2 - 3 | 921 | 83 | |
GenBank mRNA | AK041246.1 | - | 2 - 3 | 874 | 83 | |
BC138823.1 | - | 2 - 3 | 582 | 8861845 | 83 | |
M65287.1 | - | 2 - 3 | 328 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |