Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acvr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007396.4 | NP_031422.3 | 5 - 6 | 1334 | 101 | |
GenBank mRNA | AK041246.1 | - | 5 - 6 | 1287 | 101 | |
AK143186.1 | - | 6 - 7 | 1433 | 101 | ||
BC138823.1 | - | 5 - 6 | 995 | 8861845 | 101 | |
M65287.1 | - | 5 - 6 | 741 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |