Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200016693.mm.1 |
NM_001013802.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016693.mm.1 |
NM_001013802.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016693.mm.1 |
NM_028387.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016693.mm.1 |
NM_028387.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016693.mm.1 |
TC0200002099.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016694.mm.1 |
NM_001013802.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016694.mm.1 |
NM_001013802.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016694.mm.1 |
NM_028387.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016694.mm.1 |
NM_028387.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016694.mm.1 |
TC0200002099.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016705.mm.1 |
NM_001013802.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016705.mm.1 |
NM_001013802.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016705.mm.1 |
TC0200002099.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016696.mm.1 |
NM_001013802.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016696.mm.1 |
NM_001013802.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016696.mm.1 |
NM_028387.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016696.mm.1 |
NM_028387.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016696.mm.1 |
TC0200002099.mm.1 |
MTA TC |