Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cry2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009963.4 | NP_034093.1 | 3 - 4 | 504 | 72 | |
GenBank mRNA | AB003433.1 | - | 3 - 4 | 389 | 72 | |
AF156987.1 | - | 3 - 4 | 472 | 72 | ||
AK041696.1 | - | 3 - 4 | 472 | 72 | ||
AK133781.1 | - | 3 - 4 | 479 | 72 | ||
AK172994.1 | - | 3 - 4 | 436 | 72 | ||
BC054794.1 | - | 3 - 4 | 460 | 6834104 | 72 | |
BC066799.1 | - | 3 - 4 | 463 | 30103169 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |