Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK006471.1 | - | 1 - 2 | 104 | 66 | |
BC048634.1 | - | 1 - 2 | 69 | 6770610 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |