Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028193.3 | NP_082469.2 | 12 - 13 | 1636 | 82 | |
GenBank mRNA | BC037224.1 | - | 12 - 13 | 1419 | 3257364 | 82 |
BC058112.1 | - | 2 - 3 | 232 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |