Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028193.3 | NP_082469.2 | 15 - 16 | 2027 | 106 | |
GenBank mRNA | AK010890.1 | - | 2 - 3 | 276 | 106 | |
BC037224.1 | - | 15 - 16 | 1810 | 3257364 | 106 | |
BC058112.1 | - | 5 - 6 | 623 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |