Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134066.2 | NP_598827.1 | 1 - 2 | 114 | 90 | |
GenBank mRNA | AB059565.1 | - | 1 - 2 | 114 | 90 | |
AK153898.1 | - | 1 - 2 | 132 | 90 | ||
BC034259.1 | - | 1 - 2 | 111 | 4238589 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |