Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195049.1 | NP_001181978.1 | 6 - 7 | 781 | 67 | |
NM_008778.3 | NP_032804.2 | 3 - 4 | 390 | 67 | ||
GenBank mRNA | AF082297.1 | - | 2 - 3 | 304 | 67 | |
AJ496262.1 | - | 2 - 3 | 272 | 67 | ||
BC053403.1 | - | 3 - 4 | 397 | 30060082 | 67 | |
U39738.1 | - | 6 - 7 | 788 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |