Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195046.1 | NP_001181975.1 | 5 - 6 | 567 | 84 | |
NM_001195047.1 | NP_001181976.1 | 4 - 5 | 522 | 84 | ||
NM_001195048.1 | NP_001181977.1 | 5 - 6 | 677 | 84 | ||
NM_001195049.1 | NP_001181978.1 | 7 - 8 | 941 | 84 | ||
NM_008778.3 | NP_032804.2 | 4 - 5 | 550 | 84 | ||
GenBank mRNA | AF082297.1 | - | 3 - 4 | 464 | 84 | |
AJ496262.1 | - | 3 - 4 | 432 | 84 | ||
AJ496263.1 | - | 4 - 5 | 477 | 84 | ||
AK031853.1 | - | 5 - 6 | 558 | 84 | ||
AM943853.1 | - | 4 - 5 | 495 | 84 | ||
AM943854.1 | - | 5 - 6 | 540 | 84 | ||
BC053403.1 | - | 4 - 5 | 557 | 30060082 | 84 | |
U39738.1 | - | 7 - 8 | 948 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |