Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163014.1 | NP_001156486.1 | 4 - 5 | 670 | 78 | |
GenBank mRNA | BC138170.1 | - | 4 - 5 | 654 | 8861189 | 78 |
BC145172.1 | - | 4 - 5 | 654 | 9053723 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |