Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008744.2 | NP_032770.2 | 2 - 3 | 1301 | 92 | |
GenBank mRNA | AF128865.1 | - | 1 - 2 | 1036 | 92 | |
AI840271.1 | - | 1 - 2 | 175 | 92 | ||
BC141294.1 | - | 1 - 2 | 1070 | 9055906 | 92 | |
U65418.1 | - | 1 - 2 | 1017 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |