Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080848.2 | NP_543124.1 | 2 - 3 | 303 | 67 | |
GenBank mRNA | AF416510.1 | - | 2 - 3 | 308 | 67 | |
AK075937.1 | - | 2 - 3 | 246 | 67 | ||
AK144091.1 | - | 2 - 3 | 247 | 67 | ||
BC016103.1 | - | 2 - 3 | 238 | 4919781 | 67 | |
BC025801.1 | - | 2 - 3 | 280 | 5132525 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |