Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099635.1 | NP_001093105.1 | 24 - 25 | 3152 | 89 | |
GenBank mRNA | AK132623.1 | - | 9 - 10 | 1542 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |