Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200008908.mm.1 |
NM_001025360.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008908.mm.1 |
NM_001025360.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008908.mm.1 |
NM_001025361.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008908.mm.1 |
NM_001081959.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008908.mm.1 |
NM_001081959.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008908.mm.1 |
TC1200001245.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008901.mm.1 |
NM_001025360.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008901.mm.1 |
NM_001025360.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008901.mm.1 |
NM_001025361.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008901.mm.1 |
NM_001081959.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008901.mm.1 |
NM_001081959.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008901.mm.1 |
TC1200001245.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008904.mm.1 |
NM_001025360.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008904.mm.1 |
NM_001025360.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008904.mm.1 |
NM_001025361.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008904.mm.1 |
NM_001081959.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008904.mm.1 |
NM_001081959.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008904.mm.1 |
TC1200001245.mm.1 |
MTA TC |