Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163394.1 | NP_001156866.1 | 10 - 11 | 1507 | 95 | |
NM_001163396.2 | NP_001156868.1 | 11 - 12 | 1281 | 95 | ||
GenBank mRNA | AF279662.1 | - | 9 - 10 | 1018 | 95 | |
AK137823.1 | - | 10 - 11 | 1097 | 95 | ||
AK151633.1 | - | 12 - 13 | 1283 | 95 | ||
BC096017.1 | - | 10 - 11 | 1400 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |