Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clmn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040682.1 | NP_001035772.1 | 11 - 12 | 2949 | 74 | |
NM_053155.2 | NP_444385.2 | 11 - 12 | 2949 | 74 | ||
NR_104435.1 | 12 - 13 | 3030 | 74 | |||
NR_104437.1 | 12 - 13 | 3030 | 74 | |||
GenBank mRNA | AB047978.1 | - | 11 - 12 | 2942 | 74 | |
AB059644.1 | - | 11 - 12 | 2942 | 74 | ||
AB059645.1 | - | 12 - 13 | 3023 | 74 | ||
AB059647.1 | - | 12 - 13 | 3023 | 74 | ||
AK077023.1 | - | 8 - 9 | 2531 | 74 | ||
AK220261.1 | - | 3 - 4 | 928 | 74 | ||
BC141280.1 | - | 11 - 12 | 2882 | 9055892 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |